siRNA Details
| Gene Symbol: FLJ31104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102755.1 | 1, 2 | 390 | |||
| GenBank mRNA | AK055666.1 | - | 1, 2 | 237 | ||
| BC039324.1 | - | 1, 2 | 390 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500093.2 | - | 1, 2 | 390 | Chr. 5 - 55995167 - 56003649 [+] on Build GRCh38 | |

FLJ31104
Genomic Map