siRNA Details
Gene Symbol: FLJ31104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102755.1 | 2 | 1791 | |||
GenBank mRNA | AK055666.1 | - | 2 | 1638 | ||
BC039324.1 | - | 2 | 1791 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000500093.2 | - | 2 | 1791 | Chr. 5 - 55995167 - 56003649 [+] on Build GRCh38 |
