siRNA Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324218.1 | NP_001311147.1 | 3 | 2171 | ||
| NM_001324221.1 | NP_001311150.1 | 3 | 2077 | |||
| NM_001324225.1 | NP_001311154.1 | 4 | 2156 | |||
| NM_014499.3 | NP_055314.1 | 4 | 2220 | |||
| NM_198333.2 | NP_938147.1 | 2 | 2028 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443576.3 | - | 2 | 356 | Chr. 13 - 33333679 - 33335277 [-] on Build GRCh38 | |

P2RY10
Genomic Map