siRNA Details
| Gene Symbol: LOC105372897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135085.1 | 1 | 72 | |||
| GenBank mRNA | BX119711.1 | - | 1 | 72 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448988.1 | - | 1 | 72 | Chr. 1 - 209325456 - 209328532 [+] on Build GRCh38 | |

LOC105372897
Genomic Map