siRNA Details
| Gene Symbol: LOC101928140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125849.1 | 4 | 411 | |||
| GenBank mRNA | BC033773.1 | - | 4 | 422 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928140| Gene Symbol: LOC101928140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125849.1 | 4 | 411 | |||
| GenBank mRNA | BC033773.1 | - | 4 | 422 | ||