siRNA Details
Gene Symbol: TARID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109982.1 | 6 | 1660 | |||
GenBank mRNA | BC041459.1 | - | 6 | 1636 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000607033.5 | - | 6 | 1636 | Chr. 6 - 133502252 - 133888982 [-] on Build GRCh38 |
