siRNA Details
| Gene Symbol: TARID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109982.1 | 8 | 2472 | |||
| GenBank mRNA | AK093513.1 | - | 2 | 524 | ||
| BC041459.1 | - | 8 | 2448 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607033.5 | - | 8 | 2448 | Chr. 6 - 133502252 - 133888982 [-] on Build GRCh38 | |

TARID
Genomic Map