siRNA Details
| Gene Symbol: RAPGEF4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026995.1 | 4 | 3824 | |||
| GenBank mRNA | AL157450.1 | - | 4 | 3825 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435328.1 | - | 4 | 3824 | Chr. 2 - 172723186 - 172736206 [-] on Build GRCh38 | |

RAPGEF4-AS1
Genomic Map