siRNA Details
Gene Symbol: LINC01494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110238.1 | 5 | 870 | |||
GenBank mRNA | BC038542.1 | - | 5 | 869 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419511.1 | - | 5 | 869 | Chr. 2 - 218900811 - 218930636 [-] on Build GRCh38 |
