siRNA Details
Gene Symbol: LOC101928733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX538013.1 | - | 2 | 1260 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417506.1 | - | 2 | 1257 | Chr. 7 - 149398204 - 149401456 [+] on Build GRCh38 |
