siRNA Details
| Gene Symbol: LOC105371038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135177.1 | 2 | 383 | |||
| GenBank mRNA | HY055960.1 | - | 2 | 384 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455294.1 | - | 2 | 270 | Chr. 16 - 648473 - 649200 [-] on Build GRCh38 | |

LOC105371038
Genomic Map