siRNA Details
| Gene Symbol: C1orf143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG219097.1 | - | 3 | 641 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443836.5 | - | 3 | 635 | Chr. 1 - 218510096 - 218525978 [+] on Build GRCh38 | |

C1orf143
Genomic Map