siRNA Details
| Gene Symbol: LOC101927472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120622.1 | 3 | 568 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000434792.1 | - | 3 | 563 | Chr. 10 - 104323369 - 104327004 [+] on Build GRCh38 | |

LOC101927472
Genomic Map