siRNA Details
Gene Symbol: LINC01320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126404.1 | 3 | 617 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423663.5 | - | 3 | 595 | Chr. 2 - 34677579 - 34722563 [+] on Build GRCh38 |
