siRNA Details
| Gene Symbol: TBX18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125875.1 | 2 | 231 | |||
| NR_125876.1 | 2 | 197 | ||||
| GenBank mRNA | BU853872.1 | - | 231 | |||
| DB451368.1 | - | 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBX18-AS1| Gene Symbol: TBX18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125875.1 | 2 | 231 | |||
| NR_125876.1 | 2 | 197 | ||||
| GenBank mRNA | BU853872.1 | - | 231 | |||
| DB451368.1 | - | 2 | 197 | |||