siRNA Details
Gene Symbol: LINC01350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110793.1 | 4 | 468 | |||
GenBank mRNA | AF508906.1 | - | 4 | 468 | ||
AF510427.1 | - | 4 | 458 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110793.1 | 4 | 468 | |||
GenBank mRNA | AF508906.1 | - | 4 | 468 | ||
AF510427.1 | - | 4 | 458 |