siRNA Details
| Gene Symbol: LINC01350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110793.1 | 3 | 218 | |||
| GenBank mRNA | AF508906.1 | - | 3 | 218 | ||
| AF510427.1 | - | 3 | 208 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01350| Gene Symbol: LINC01350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110793.1 | 3 | 218 | |||
| GenBank mRNA | AF508906.1 | - | 3 | 218 | ||
| AF510427.1 | - | 3 | 208 | |||