siRNA Details
| Gene Symbol: MIR646HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046099.1 | 5 | 1027 | |||
| GenBank mRNA | AK092805.1 | - | 5 | 1027 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432910.5 | - | 5 | 1027 | Chr. 20 - 60138492 - 60322256 [+] on Build GRCh38 | |

MIR646HG
Genomic Map