siRNA Details
| Gene Symbol: CELSR3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111921.1 | 1, 2 | 43 | |||
| GenBank mRNA | BM714397.1 | - | 1, 2 | 35 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421275.1 | - | 1, 2 | 35 | Chr. 3 - 48663776 - 48669174 [+] on Build GRCh38 | |

CELSR3-AS1
Genomic Map