siRNA Details
Gene Symbol: CELSR3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111921.1 | 2 | 111 | |||
GenBank mRNA | BM714397.1 | - | 2 | 103 | ||
HY035766.1 | - | 3 | 293 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000421275.1 | - | 2 | 103 | Chr. 3 - 48663776 - 48669174 [+] on Build GRCh38 |
