siRNA Details
| Gene Symbol: LOC101929116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121584.1 | 3 | 675 | |||
| GenBank mRNA | BC031239.1 | - | 3 | 677 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419853.1 | - | 3 | 678 | Chr. 9 - 126270121 - 126275918 [-] on Build GRCh38 | |

LOC101929116
Genomic Map