siRNA Details
Gene Symbol: HSPC324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135132.1 | 2 | 448 | |||
GenBank mRNA | AF161442.1 | - | 2 | 448 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000411904.1 | - | 2 | 448 | Chr. 9 - 136648610 - 136660421 [-] on Build GRCh38 |
