siRNA Details
Gene Symbol: LOC100506885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134649.1 | 3 | 238 | |||
NR_134650.1 | 2 | 130 | ||||
GenBank mRNA | AA810378.1 | - | 2 | 120 | ||
AK092541.1 | - | 3 | 229 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134649.1 | 3 | 238 | |||
NR_134650.1 | 2 | 130 | ||||
GenBank mRNA | AA810378.1 | - | 2 | 120 | ||
AK092541.1 | - | 3 | 229 |