siRNA Details
| Gene Symbol: LINC00687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110635.1 | 1 | 15 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557899.1 | - | 1 | 15 | Chr. 20 - 11810021 - 11870715 [-] on Build GRCh38 | |

LINC00687
Genomic Map