siRNA Details
| Gene Symbol: LOC105378663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135048.1 | 2 | 392 | |||
| GenBank mRNA | BX115242.1 | - | 2 | 343 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443161.1 | - | 2 | 351 | Chr. 1 - 38873442 - 38876226 [+] on Build GRCh38 | |

LOC105378663
Genomic Map