siRNA Details
| Gene Symbol: LOC105378663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135048.1 | 2 | 420 | |||
| GenBank mRNA | BX115242.1 | - | 2 | 371 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378663| Gene Symbol: LOC105378663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135048.1 | 2 | 420 | |||
| GenBank mRNA | BX115242.1 | - | 2 | 371 | ||