siRNA Details
| Gene Symbol: TSTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195131.1 | NP_001182060.1 | 4 | 452 | ||
| XM_017010141.1 | XP_016865630.1 | 4 | 1343 | |||
| GenBank mRNA | BC047952.1 | - | 3 | 301 | ||
| BX647075.1 | - | 5 | 538 | |||
| DN996863.1 | - | 4 | 416 | |||

TSTD3
Genomic Map