siRNA Details
Gene Symbol: LINC01546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038428.1 | 3 | 479 | |||
GenBank mRNA | BX101503.1 | - | 3 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457435.1 | - | 3 | 479 | Chr. X - 3271820 - 3284653 [+] on Build GRCh38 |
