siRNA Details
| Gene Symbol: LINC01546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038428.1 | 3 | 243 | |||
| GenBank mRNA | BX101503.1 | - | 3 | 243 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457435.1 | - | 3 | 243 | Chr. X - 3271820 - 3284653 [+] on Build GRCh38 | |

LINC01546
Genomic Map