siRNA Details
Gene Symbol: LOC101927692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120628.1 | 3 | 1439 | |||
GenBank mRNA | AK098198.1 | - | 3 | 1433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000436932.1 | - | 3 | 1439 | Chr. 10 - 114764788 - 114779903 [+] on Build GRCh38 |
