siRNA Details
| Gene Symbol: LOC101927692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120628.1 | 3 | 1302 | |||
| GenBank mRNA | AK098198.1 | - | 3 | 1296 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436932.1 | - | 3 | 1302 | Chr. 10 - 114764788 - 114779903 [+] on Build GRCh38 | |

LOC101927692
Genomic Map