siRNA Details
Gene Symbol: LINC01013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040870.1 | - | 2 | 889 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000458028.1 | - | 2 | 889 | Chr. 6 - 132133978 - 132147058 [+] on Build GRCh38 |
