siRNA Details
| Gene Symbol: LOC440570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135765.1 | 1 | 573 | |||
| GenBank mRNA | AK125737.1 | - | 1 | 574 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000438002.1 | - | 1 | 574 | Chr. 1 - 16870945 - 16874079 [+] on Build GRCh38 | |

LOC440570
Genomic Map