siRNA Details
| Gene Symbol: D21S2088E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040254.1 | 5 | 928 | |||
| GenBank mRNA | AY063451.1 | - | 5 | 928 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000262354.4 | - | 954 | Chr. 9 - 5456114 - 5467862 [+] on Build GRCh38 | ||

D21S2088E
Genomic Map