siRNA Details
| Gene Symbol: DCAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026213.1 | - | 1 | 956 | ||
| Gene Symbol: LOC100287049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA104933.1 | - | 2 | 556 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000442130.1 | - | 2 | 471 | Chr. 1 - 160261744 - 160262778 [+] on Build GRCh38 | |

DCAF8
Genomic Map