siRNA Details
Gene Symbol: ZNF883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101338.1 | NP_001094808.1 | 5 | 397 | ||
GenBank mRNA | AK095843.1 | - | 5 | 397 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101338.1 | NP_001094808.1 | 5 | 397 | ||
GenBank mRNA | AK095843.1 | - | 5 | 397 |