siRNA Details
Gene Symbol: LOC101927768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125845.1 | 3 | 811 | |||
GenBank mRNA | BC042098.1 | - | 3 | 811 | ||
CD643945.1 | - | 3 | 543 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000519104.5 | - | 3 | 811 | Chr. 6 - 113969701 - 114340738 [+] on Build GRCh38 |
