siRNA Details
| Gene Symbol: LOC101927768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125845.1 | 1 | 21 | |||
| GenBank mRNA | BC042098.1 | - | 1 | 21 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000519104.5 | - | 1 | 21 | Chr. 6 - 113969701 - 114340738 [+] on Build GRCh38 | |

LOC101927768
Genomic Map