siRNA Details
| Gene Symbol: LINC00448 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047029.1 | 8 | 728 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448411.1 | - | 4 | 363 | Chr. 13 - 62672285 - 62732180 [-] on Build GRCh38 | |

LINC00448
Genomic Map