siRNA Details
| Gene Symbol: LOC101927416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121663.1 | 1 | 187 | |||
| NR_121664.1 | 1 | 187 | ||||
| GenBank mRNA | BC041457.1 | - | 1 | 145 | ||
| DB198681.1 | - | 1 | 187 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927416| Gene Symbol: LOC101927416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121663.1 | 1 | 187 | |||
| NR_121664.1 | 1 | 187 | ||||
| GenBank mRNA | BC041457.1 | - | 1 | 145 | ||
| DB198681.1 | - | 1 | 187 | |||