siRNA Details
| Gene Symbol: SPATA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104595.1 | 5 | 1439 | |||
| GenBank mRNA | BC043582.1 | - | 5 | 1253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439928.2 | - | 5 | 1255 | Chr. 13 - 23979810 - 24035027 [+] on Build GRCh38 | |

SPATA13
Genomic Map