siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047526.1 | 3 | 652 | |||
| GenBank mRNA | BC086872.1 | - | 2 | 438 | ||
| BX095144.1 | - | 3 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01128| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047526.1 | 3 | 652 | |||
| GenBank mRNA | BC086872.1 | - | 2 | 438 | ||
| BX095144.1 | - | 3 | 524 | |||