siRNA Details
| Gene Symbol: LAMA5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109922.1 | 1 | 139 | |||
| NR_109923.1 | 1 | 139 | ||||
| NR_109924.1 | 1 | 139 | ||||
| GenBank mRNA | BM700151.1 | - | 1 | 139 | ||
| BQ188912.1 | - | 1 | 138 | |||
| CD675432.1 | - | 1 | 137 | |||

LAMA5-AS1
Genomic Map