siRNA Details
| Gene Symbol: MIR4500HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033829.1 | 6 | 1535 | |||
| GenBank mRNA | AF339814.1 | - | 6 | 1549 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436290.2 | - | 6 | 1535 | Chr. 13 - 87443987 - 87670963 [-] on Build GRCh38 | |

MIR4500HG
Genomic Map