siRNA Details
| Gene Symbol: SATB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125803.1 | 4, 5 | 409 | |||
| GenBank mRNA | AW027730.1 | - | 4, 5 | 373 | ||
| BF115425.1 | - | 4, 5 | 410 | |||
| DR006704.1 | - | 4, 5 | 628 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414198.5 | - | 4, 5 | 622 | Chr. 3 - 18445024 - 18526508 [+] on Build GRCh38 | |

SATB1-AS1
Genomic Map