siRNA Details
| Gene Symbol: LOC102724502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135522.1 | 2 | 451 | |||
| GenBank mRNA | BX095520.1 | - | 2 | 451 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000424837.1 | - | 2 | 451 | Chr. 21 - 32958888 - 32960566 [+] on Build GRCh38 | |

LOC102724502
Genomic Map