siRNA Details
| Gene Symbol: LOC100996635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110621.1 | 3 | 928 | |||
| NR_110622.1 | 2 | 825 | ||||
| GenBank mRNA | BC045746.2 | - | 2 | 681 | ||
| BC067883.1 | - | 3 | 703 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000411798.1 | - | 3 | 704 | Chr. 1 - 95510340 - 95515464 [+] on Build GRCh38 | |

LOC100996635
Genomic Map