siRNA Details
| Gene Symbol: LOC101927159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110004.1 | 4 | 1218 | |||
| GenBank mRNA | BC040166.1 | - | 4 | 1218 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000457704.2 | - | 4 | 1218 | Chr. 20 - 42685483 - 42688562 [+] on Build GRCh38 | |

LOC101927159
Genomic Map