siRNA Details
Gene Symbol: LOC101927244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125938.1 | 7 | 1490 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000419450.1 | - | 7 | 1490 | Chr. 1 - 70706453 - 70786468 [+] on Build GRCh38 |
