siRNA Details
Gene Symbol: LOC101927482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110225.1 | 3 | 915 | |||
NR_110226.1 | 3 | 904 | ||||
GenBank mRNA | AK023739.1 | - | 2 | 904 | ||
BC018675.1 | - | 2 | 794 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433933.1 | - | 2 | 904 | Chr. 2 - 196260114 - 196264204 [+] on Build GRCh38 |
